1-Butylimidazole 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Butyl-1H-imidazole, 1-Butyl-1H-imidazole, N-Butylimidazole

L07793-14 L07793-22
CAAAAL07793-14EA 71.73 CAD
CAAAAL07793-14 CAAAAL07793-22
1-Butylimidazole 99%
1-Butylimidazole

Purity: 99% CAS No.: 4316-42-1 Molecular Formula: C7H12N2 Formula Weight: 124.19 Flash Point: 145°(293°F) Refractive Index: 1.4800 Odor: Amine-like Boiling Point: 257° Beilstein Reference: 109499 UN No.: 2690 MDL No.: MFCD00042753

Formula: C₇H₁₂N₂
MW: 124.19 g/mol
Boiling Pt: 257 °C
Density: 0.948
Flash Pt: 145 °C (293 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042753
CAS Number: 4316-42-1
EINECS: 224-335-9
UN: 2690
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR