2,5-Dichlorobenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (2,5-Dichlorophenyl)methanol

L07734-06 L07734-14
CAAAAL07734-06EA 63.2 CAD
CAAAAL07734-06 CAAAAL07734-14
2,5-Dichlorobenzyl alcohol 99%
2,5-Dichlorobenzyl alcohol

Purity: 99% CAS No.: 34145-05-6 Molecular Formula: C7H6Cl2O Formula Weight: 177.03 Melting Point: 76-80° Beilstein Reference: 3240770

Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 264
Melting Pt: 80
Storage Temperature: Ambient
MDL Number: MFCD00004607
CAS Number: 34145-05-6
EINECS: 251-850-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR