D(-)-Lysine monohydrochloride 98%
Supplier: Thermo Scientific Chemicals
L07710-03
L07710-06
CAAAAL07710-03EA
60.67
CAD
CAAAAL07710-03
CAAAAL07710-06
D(-)-Lysine monohydrochloride 98%
D(-)-Lysine monohydrochloride
Purity: 98%
CAS No.: 7274-88-6
Molecular Formula: C6H14N2O2.HCl
Formula Weight: 182.65
Melting Point: ca 260° dec.
Beilstein Reference: 4356907
Formula:
C₆H₁₅ClN₂O₂ Melting Pt: ∼ 260 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012920 CAS Number: 7274-88-6 EINECS: 230-691-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...