Borane dimethyl sulfide complex 94%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L07705-AC
L07705-AE
CAAAAL07705-ACEA
104.6
CAD
CAAAAL07705-AC
CAAAAL07705-AE
Borane dimethyl sulfide complex 94%
Borane dimethyl sulphide complex
Purity: 94%
CAS No.: 13292-87-0
Molecular Formula: C2H9BS
Formula Weight: 75.97
Storage Temperature: 2°C to 8°C
Flash Point: -1°(30°F)
Refractive Index: 1.4570
Melting Point: -38°
Boiling Point: ca 44° dec.
Beilstein Reference: 3663489
UN No.: 2924
MDL No.: MFCD00013189
Formula:
C₂H₉BS Boiling Pt: ∼ 44 °C (dec.) Melting Pt: –38 °C Density: 0.801 Flash Pt: –1 °C (30 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013189 CAS Number: 13292-87-0 EINECS: 236-313-6 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...