Chloroacetaldehyde diethyl acetal 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L07661-22
L07661-36
CAAAAL07661-22EA
81.05
CAD
CAAAAL07661-22
CAAAAL07661-36
Chloroacetaldehyde diethyl acetal 98%
Chloroacetaldehyde diethyl acetal
Purity: 98%
CAS No.: 621-62-5
Molecular Formula: C6H13ClO2
Formula Weight: 152.62
Flash Point: 50°(122°F)
Refractive Index: 1.4160
Melting Point: -45°
Boiling Point: 156-157°
Beilstein Reference: 969254
UN No.: 1993
MDL No.: MFCD00000949
Formula:
C₆H₁₃ClO₂ Boiling Pt: 156…157 °C Melting Pt: –45 °C Density: 1.018 Flash Pt: 50 °C (122 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000949 CAS Number: 621-62-5 EINECS: 210-695-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...