Ethyl-2-iodobenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

L07650-06 L07650-14
CAAAAL07650-06EA 81.21 CAD
CAAAAL07650-06 CAAAAL07650-14
Ethyl-2-iodobenzoate 98%
Formula: C₉H₉IO₂
Boiling Pt: 163…165 °C/23 mm
Storage Temperature: Ambient
MDL Number: MFCD00051796
CAS Number: 1829-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR