Dibutyl sulfide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L07599-AE
L07599-AP
CAAAAL07599-AEEA
96.7
CAD
CAAAAL07599-AE
CAAAAL07599-AP
Dibutyl sulfide 98%
Dibutyl sulphide
Purity: 98%
CAS No.: 544-40-1
Molecular Formula: C8H18S
Formula Weight: 146.30
Flash Point: 76°(168°F)
Refractive Index: 1.4530
Odor: Stench
Melting Point: -80°
Boiling Point: 188-189°
Beilstein Reference: 1732829
Merck Reference: 14,1590
UN No.: 3334
MDL No.: MFCD00009468
Formula:
C₈H₁₈S Boiling Pt: 188…189 °C Melting Pt: –80 °C Density: 0.839 Flash Pt: 76 °C (168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009468 CAS Number: 544-40-1 EINECS: 208-870-5 UN: 3334 ADR: 9, Merck Index: 14,1590 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...