2-Methyl-3-butyne-2-ol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L07593-0F
L07593-AE
L07593-AP
CAAAAL07593-0FEA
301.78
CAD
CAAAAL07593-0F
CAAAAL07593-AE
CAAAAL07593-AP
2-Methyl-3-butyne-2-ol 98%
2-Methyl-3-butyne-2-ol
Purity: 98%
CAS No.: 115-19-5
Molecular Formula: C5H8O
Formula Weight: 84.12
Flash Point: 22°(72°F)
Refractive Index: 1.4210
Melting Point: 2-3°
Boiling Point: 103-104°
Beilstein Reference: 635746
Merck Reference: 14,6034
UN No.: 1987
MDL No.: MFCD00004467
Formula:
C₅H₈O Boiling Pt: 103…104 °C Melting Pt: 2…3 °C Density: 0.863 Flash Pt: 22 °C (72 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004467 CAS Number: 115-19-5 EINECS: 204-070-5 UN: 1987 ADR: 3,III Merck Index: 14,6034 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...