DL-Lysine monohydrate 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
(±)-Lysine monohydrate, (±)-2,6-Diaminohexanoic acid monohydrate, (±)-2,6-Diaminocaproic acid monohydrate, H-DL-Lys-OH monohydrate
L07576-06
L07576-14
CAAAAL07576-06EA
55.62
CAD
CAAAAL07576-06
CAAAAL07576-14
DL-Lysine monohydrate 98+%
DL-Lysine monohydrate
Purity: 98+%
CAS No.: 885701-25-7
Molecular Formula: C6H14N2O2.H2O
Formula Weight: 164.21 (146.19anhy)
Melting Point: ca 110°
Beilstein Reference: 1616991
Formula:
C₆H₁₄N₂O₂·1H₂O Melting Pt: ∼ 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151036 CAS Number: 885701-25-7 EINECS: 200-740-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...