1-Bromo-2-chloro-4-nitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

L07573-09 L07573-18
CAAAAL07573-09EA 96.06 CAD
CAAAAL07573-09 CAAAAL07573-18
1-Bromo-2-chloro-4-nitrobenzene 98%
1-Bromo-2-chloro-4-nitrobenzene

Purity: 98% CAS No.: 29682-39-1 Molecular Formula: C6H3BrClNO2 Formula Weight: 236.45 Melting Point: 59-62° Beilstein Reference: 2253107 UN No.: 2811 MDL No.: MFCD00051514

Formula: C₆H₃BrClNO₂
MW: 236.45 g/mol
Melting Pt: 62
Storage Temperature: Ambient
MDL Number: MFCD00051514
CAS Number: 29682-39-1
EINECS: 249-773-8
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR