5-Chloro-3-hydroxypyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chloro-5-pyridinol, 3-Chloro-5-hydroxypyridine, 5-Chloro-3-pyridinol

L07568-06 L07568-14
CAAAAL07568-06EA 81.37 CAD
CAAAAL07568-06 CAAAAL07568-14
5-Chloro-3-hydroxypyridine 99%
5-Chloro-3-hydroxypyridine

Purity: 99% CAS No.: 74115-12-1 Molecular Formula: C5H4ClNO Formula Weight: 129.55 Melting Point: 159-163° Beilstein Reference: 1422550

Formula: C₅H₄ClNO
MW: 129.55 g/mol
Melting Pt: 159…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00006379
CAS Number: 74115-12-1
EINECS: 277-727-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR