Ethidium bromide 98% (dry weight)
Supplier: Thermo Scientific Chemicals
|
Danger
|
L07482-03
L07482-06
CAAAAL07482-03EA
59.09
CAD
CAAAAL07482-03
CAAAAL07482-06
Ethidium bromide 98% (dry weight)
Ethidium bromide
For DNA isolation and visualization
Purity: 98%
CAS No.: 1239-45-8
Molecular Formula: C21H20BrN3
Formula Weight: 394.32
Odor: Weak
Melting Point: ca 261° dec.
Beilstein Reference: 3642536
Merck Reference: 14,4731
UN No.: 2811
MDL No.: MFCD00011724
Formula:
C₂₁H₂₀BrN₃ Boiling Pt: 72 °C (0.1 -rr) Melting Pt: ∼ 261 °C (dec.) Density: 1.01 g/cm³ (20 °C) Flash Pt: ≤ 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011724 CAS Number: 1239-45-8 EINECS: 214-984-6 UN: 2811 ADR: 6.1,III Merck Index: 14,4731 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...