Methyl linoleate 95%
Supplier: Thermo Scientific Chemicals
L07450-03
L07450-06
CAAAAL07450-03EA
53.4
CAD
CAAAAL07450-03
CAAAAL07450-06
Methyl linoleate 95%
Methyl linoleate
Purity: 95%
CAS No.: 112-63-0
Molecular Formula: C19H34O2
Formula Weight: 294.47
Storage Temperature: 2°C to 8°C
Flash Point: 200°(392°F)
Refractive Index: 1.4640
Melting Point: -35°
Boiling Point: 207-208°/11mm
Beilstein Reference: 1727614
Merck Reference: 14,6095
MDL No.: MFCD00009534
Formula:
C₁₉H₃₄O₂ Boiling Pt: 207…208 °C/11 mm Melting Pt: –35 °C Density: 0.888 Flash Pt: 200 °C(392 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009534 CAS Number: 112-63-0 EINECS: 203-993-0 Merck Index: 14,6095 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...