Methyl-4-chloro-3-nitrobenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L07407-06
L07407-14
CAAAAL07407-06EA
91.32
CAD
CAAAAL07407-06
CAAAAL07407-14
Methyl-4-chloro-3-nitrobenzoate 98%
Methyl 4-chloro-3-nitrobenzoate
Purity: 98%
CAS No.: 14719-83-6
Molecular Formula: C8H6ClNO4
Formula Weight: 215.59
Melting Point: 78-82°
Beilstein Reference: 1643729
Formula:
C₈H₆ClNO₄ Melting Pt: 78…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016989 CAS Number: 14719-83-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...