2-Bromononane 97%

Supplier: Thermo Scientific Chemicals

L07387-03 L07387-06
CAAAAL07387-03EA 57.99 CAD
CAAAAL07387-03 CAAAAL07387-06
2-Bromononane 97%
2-Bromononane

Purity: 97% CAS No.: 2216-35-5 Molecular Formula: C9H19Br Formula Weight: 207.16 Refractive Index: 1.4530 Boiling Point: 208-209° MDL No.: MFCD00017865

Formula: H₃CBrCH(CH₂)₆CH₃
Boiling Pt: 208…209 °C
Density: 1.081
Storage Temperature: Ambient
MDL Number: MFCD00017865
CAS Number: 2216-35-5
EINECS: 218-688-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR