2-Isopropoxyphenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L07360-06
L07360-14
CAAAAL07360-06EA
65.41
CAD
CAAAAL07360-06
CAAAAL07360-14
2-Isopropoxyphenol 97%
2-Isopropoxyphenol
Purity: 97%
CAS No.: 4812-20-8
Molecular Formula: C9H12O2
Formula Weight: 152.19
Flash Point: 87°(188°F)
Refractive Index: 1.5150
Boiling Point: 100-102°/11mm
Beilstein Reference: 1937063
MDL No.: MFCD00002184
Formula:
C₉H₁₂O₂ Boiling Pt: 100…102 °C (11 mmHg) Melting Pt: 206…207 °C Density: 1,032 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002184 CAS Number: 4812-20-8 EINECS: 225-379-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...