Trimellitic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2,4-Benzenetricarboxylic acid, Benzene-1,2,4-tricarboxylic acid

L07359-14 L07359-22
CAAAAL07359-14EA 84.21 CAD
CAAAAL07359-14 CAAAAL07359-22
Trimellitic acid 98%
Trimellitic acid

Purity: 98% CAS No.: 528-44-9 Molecular Formula: C9H6O6 Formula Weight: 210.14 Melting Point: ca 231° dec. Beilstein Reference: 2214815 Merck Reference: 14,9702 MDL No.: MFCD00002470

Formula: C₉H₆O₆
MW: 210.14 g/mol
Boiling Pt: 107 - 113 °C (0.1 -rr)
Melting Pt: ca 231 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002470
CAS Number: 528-44-9
EINECS: 208-432-3
Merck Index: 14,9702

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR