(±)-2-Methyltetrahydrofuran 99% stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) ∼ 150-400 ppm

L07356-AE L07356-AP L07356-0F
CAAAAL07356-AEEA 69.84 CAD
CAAAAL07356-AE CAAAAL07356-AP CAAAL07356-0F
(±)-2-Methyltetrahydrofuran 99% stabilized
(±)-2-Methyltetrahydrofuran

Purity: 99% CAS No.: 96-47-9 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: -11°(12°F) Refractive Index: 1.4060 Melting Point: -136° Boiling Point: 80-82° Beilstein Reference: 102448 UN No.: 2536 MDL No.: MFCD00005367

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 80…82 °C
Melting Pt: –136 °C
Density: 0.856
Flash Pt: -11 °C(12 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005367
CAS Number: 96-47-9
EINECS: 202-507-4
UN: 2536
ADR: "3",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR