1,4-Cyclohexadiene 97% stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 0.01%

L07337-06 L07337-14 L07337-22
CAAAAL07337-06EA 72.05 CAD
CAAAAL07337-06 CAAAAL07337-14 CAAAAL07337-22
1,4-Cyclohexadiene 97% stabilized
1,4-Cyclohexadiene

Purity: 97% CAS No.: 628-41-1 Molecular Formula: C6H8 Formula Weight: 80.13 Storage Temperature: 2°C to 8°C Flash Point: -6°(21°F) Refractive Index: 1.4720 Melting Point: -49° Boiling Point: 88-90° Beilstein Reference: 1900733 UN No.: 3295 MDL No.: MFCD00001535

Formula: C₆H₈
Boiling Pt: 88…90 °C
Melting Pt: –49 °C
Density: 0.847
Flash Pt: –6 °C (21 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001535
CAS Number: 628-41-1
EINECS: 211-043-1
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR