4-Fluorobenzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Fluorobenzene-1-sulfonyl chloride, 4-Fluorobenzene-1-sulphonyl chloride

L07299-14 L07299-22
CAAAAL07299-14EA 100.96 CAD
CAAAAL07299-14 CAAAAL07299-22
4-Fluorobenzenesulfonyl chloride 98%
4-Fluorobenzenesulphonyl chloride

Purity: 98% CAS No.: 349-88-2 Molecular Formula: C6H4ClFO2S Formula Weight: 194.61 Flash Point: 110°(230°F) Melting Point: 35-38° Boiling Point: 86-87°/1mm Beilstein Reference: 2091633 UN No.: 3261 MDL No.: MFCD00007438

Formula: C₆H₄ClFO₂S
MW: 194.61 g/mol
Boiling Pt: 86…87 °C (1 mmHg)
Melting Pt: 35…38 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007438
CAS Number: 349-88-2
EINECS: 206-493-0
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR