2-tert-Butylphenol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L07294-0P
L07294-22
CAAAAL07294-0PEA
176.96
CAD
CAAAAL07294-0P
CAAAAL07294-22
2-tert-Butylphenol 99%
2-tert-Butylphenol
Purity: 99%
CAS No.: 88-18-6
Molecular Formula: C10H14O
Formula Weight: 150.22
Flash Point: 102°(215°F)
Refractive Index: 1.5230
Melting Point: -7°
Boiling Point: 223-224°
Beilstein Reference: 1907120
UN No.: 2922
MDL No.: MFCD00002223
Formula:
C₁₀H₁₄O Boiling Pt: 223…224 °C Melting Pt: –7 °C Density: 0.980 Flash Pt: 102 °C (215 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002223 CAS Number: 88-18-6 EINECS: 201-807-2 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...