2-Ethyl-9,10-anthraquinone 98%

Supplier: Thermo Scientific Chemicals

L07206-06 L07206-22 L07206-36
CAAAAL07206-06EA 48.98 CAD
CAAAAL07206-06 CAAAAL07206-22 CAAAAL07206-36
2-Ethyl-9,10-anthraquinone 98%
2-Ethyl-9,10-anthraquinone

Purity: 98% CAS No.: 84-51-5 Molecular Formula: C16H12O2 Formula Weight: 236.27 Flash Point: 210°(410°F) Melting Point: 107-111° Boiling Point: 180-190° Beilstein Reference: 1969873 MDL No.: MFCD00001237

Formula: C₁₆H₁₂O₂
Boiling Pt: 180…190 °C
Melting Pt: 107…111 °C
Flash Pt: >210 °C (410 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001237
CAS Number: 84-51-5
EINECS: 201-535-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR