1,9-Nonanedithiol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,9-Dimercaptononane
L07172-03
L07172-06
CAAAAL07172-03EA
66.68
CAD
CAAAAL07172-03
CAAAAL07172-06
1,9-Nonanedithiol 97%
1,9-Nonanedithiol
Purity: 97%
CAS No.: 3489-28-9
Molecular Formula: HS(CH2)9SH
Formula Weight: 192.39
Flash Point:
Formula:
C₉H₂₀S₂ MW: 192.39 g/mol Boiling Pt: 284 Melting Pt: -17.5 Density: 0.953 Flash Pt: >100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004914 CAS Number: 3489-28-9 EINECS: 222-482-3 UN: 3334 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...