4-Fluorobiphenyl 97+%

Supplier: Thermo Scientific Chemicals
Warning

L07170-03 L07170-06
CAAAAL07170-03EA 57.51 CAD
CAAAAL07170-03 CAAAAL07170-06
4-Fluorobiphenyl 97+%
4-Fluorobiphenyl

Purity: 97+% CAS No.: 324-74-3 Molecular Formula: C12H9F Formula Weight: 172.20 Melting Point: 72-76° Boiling Point: 253° MDL No.: MFCD00011650

Formula: C₁₂H₉F
Boiling Pt: 253 °C
Melting Pt: 72…76 °C
Density: 1.247 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011650
CAS Number: 324-74-3
EINECS: 206-304-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR