(S)-(+)-Carvone 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: D-p-Mentha-1(6),8-dien-2-one, D(+)-Carvone, (S)-2-Methyl-5-(prop-1-en-2-yl)cyclohex-2-enone, (S)-(+)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-one

L07130-14 L07130-22
CAAAAL07130-14EA 98.91 CAD
CAAAAL07130-14 CAAAAL07130-22
(S)-(+)-Carvone 96%
(S)-(+)-Carvone

Purity: 96% CAS No.: 2244-16-8 Molecular Formula: C10H14O Formula Weight: 150.22 Flash Point: 93°(199°F) Refractive Index: 1.4980 Odor: Pleasant Boiling Point: 228-230° Beilstein Reference: 2042970 Merck Reference: 14,1874 Optical Rotation: +59° (neat) MDL No.: MFCD00062997

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 228…230 °C
Melting Pt: ? 15 °C
Density: 0.959
Flash Pt: 93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00062997
CAS Number: 2244-16-8
EINECS: 218-827-2
Merck Index: 14,1874

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR