2-Biphenylylmethanol 98%
Supplier: Thermo Scientific Chemicals
L07094-03
L07094-06
CAAAAL07094-03EA
45.35
CAD
CAAAAL07094-03
CAAAAL07094-06
2-Biphenylylmethanol 98%
2-Biphenylyl-methanol
Purity: 98%
CAS No.: 2928-43-0
Molecular Formula: C13H12O
Formula Weight: 184.24
Melting Point: 47-51°
Boiling Point: 96°/0.04mm
MDL No.: MFCD00004620
Formula:
C₁₃H₁₂O Boiling Pt: 96 °C/0.04 mm Melting Pt: 47…51 °C Density: 1,2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004620 CAS Number: 2928-43-0 EINECS: 220-892-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...