(±)-trans-1,2-Cyclohexanediol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (±)-trans-Cyclohexane-1,2-diol, trans-Cyclohexane-1,2-diol

L07088-14 L07088-22
CAAAAL07088-14EA 118.5 CAD
CAAAAL07088-14 CAAAAL07088-22
(±)-trans-1,2-Cyclohexanediol 98%
(±)-trans-1,2-Cyclohexanediol

Purity: 98% CAS No.: 1460-57-7 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 134°(273°F) Melting Point: 101-106° Boiling Point: 231-233° Beilstein Reference: 3193810 MDL No.: MFCD00063611

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 231…233 °C
Melting Pt: 101…106 °C
Density: 1.147 g/cm³ (24 °C)
Flash Pt: 134 °C(273 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063611
CAS Number: 1460-57-7
EINECS: 215-956-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR