1-Heptanethiol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
n-Heptanethiol, n-Heptyl mercaptan, 1-Mercaptoheptane
L07079-14
L07079-22
CAAAAL07079-14EA
83.9
CAD
CAAAAL07079-14
CAAAAL07079-22
1-Heptanethiol 98%
1-Heptanethiol
Purity: 98%
CAS No.: 1639-09-4
Molecular Formula: CH3(CH2)6SH
Formula Weight: 132.27
Flash Point: 46°(114°F)
Refractive Index: 1.4515
Melting Point: -43°
Boiling Point: 173-175°
Beilstein Reference: 1731687
UN No.: 3336
MDL No.: MFCD00004911
Formula:
C₇H₁₆S Boiling Pt: 173…175 °C Melting Pt: –43 °C Density: 0.854 Flash Pt: 46 °C (114 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004911 CAS Number: 1639-09-4 EINECS: 216-678-8 UN: 3336 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...