3-Hydroxy-2-nitro-6-picoline 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Methyl-2-nitropyridin-3-ol, 6-Methyl-2-nitro-3-pyridone, 3-Hydroxy-6-methyl-2-nitropyridine, 6-Metil-2-nitropyridine-3-ol

L07074-09 L07074-18
CAAAAL07074-09EA 157.84 CAD
CAAAAL07074-09 CAAAAL07074-18
3-Hydroxy-2-nitro-6-picoline 99%
3-Hydroxy-2-nitro-6-picoline

Purity: 99% CAS No.: 15128-90-2 Molecular Formula: C6H6N2O3 Formula Weight: 154.13 Melting Point: 105-108° Beilstein Reference: 974751

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00006259
CAS Number: 15128-90-2
EINECS: 239-192-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR