3-Bromo-5-nitrosalicylaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L07071-03
L07071-06
L07071-14
CAAAAL07071-03EA
42.82
CAD
CAAAAL07071-03
CAAAAL07071-06
CAAAAL07071-14
3-Bromo-5-nitrosalicylaldehyde 98%
3-Bromo-5-nitrosalicylaldehyde
Purity: 98%
CAS No.: 16789-84-7
Molecular Formula: C7H4BrNO4
Formula Weight: 246.02
Melting Point: 147-151°
Beilstein Reference: 1213613
Formula:
C₇H₄BrNO₄ Melting Pt: 147…151 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051833 CAS Number: 16789-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...