3-Methoxypropiononitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methoxypropanenitrile, 3-Methoxypropionitrile

L07055-AE L07055-AP
CAAAAL07055-AEEA 45.66 CAD
CAAAAL07055-AE CAAAAL07055-AP
3-Methoxypropiononitrile 99%
3-Methoxypropiononitrile

Purity: 99% CAS No.: 110-67-8 Molecular Formula: CH3O(CH2)2CN Formula Weight: 85.11 Flash Point: 66°(150°F) Refractive Index: 1.4030 Odor: Weak pH: 6.8 Melting Point: -63° Boiling Point: 164-165° Beilstein Reference: 1739284 MDL No.: MFCD00001958

Formula: C₄H₇NO
MW: 85.11 g/mol
Boiling Pt: 164…165 °C
Melting Pt: –63 °C
Density: 0,938
Flash Pt: 66 °C(150 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001958
CAS Number: 110-67-8
EINECS: 203-790-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR