2-Iodobiphenyl 98%

Supplier: Thermo Scientific Chemicals
Warning

L06990-03 L06990-06
CAAAAL06990-03EA 65.1 CAD
CAAAAL06990-03 CAAAAL06990-06
2-Iodobiphenyl 98%
2-Iodobiphenyl

Purity: 98% CAS No.: 2113-51-1 Molecular Formula: C12H9I Formula Weight: 280.11 Flash Point: 110°(230°F) Refractive Index: 1.6610 Boiling Point: 300° Beilstein Reference: 2206820 MDL No.: MFCD00039396

Formula: C₁₂H₉I
Boiling Pt: 300 °C
Density: 1.590
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039396
CAS Number: 2113-51-1
EINECS: 218-303-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR