2-Methyl-8-quinolinol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06989-22
L06989-36
CAAAAL06989-22EA
89.9
CAD
CAAAAL06989-22
CAAAAL06989-36
2-Methyl-8-quinolinol 98%
2-Methyl-8-quinolinol
Purity: 98%
CAS No.: 826-81-3
Molecular Formula: C10H9NO
Formula Weight: 159.19
Melting Point: 70-74°
Boiling Point: 267°
UN No.: 3077
MDL No.: MFCD00006765
Formula:
C₁₀H₉NO Boiling Pt: 267 °C Melting Pt: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006765 CAS Number: 826-81-3 EINECS: 212-562-6 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...