(Chloromethyl)cyclopropane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cyclopropyl methyl chloride
L06986-06
L06986-14
CAAAAL06986-06EA
145.68
CAD
CAAAAL06986-06
CAAAAL06986-14
(Chloromethyl)cyclopropane 97%
(Chloromethyl)cyclopropane
Purity: 97%
CAS No.: 5911-08-0
Molecular Formula: C4H7Cl
Formula Weight: 90.55
Storage Temperature: 2°C to 8°C
Flash Point: -1°(30°F)
Refractive Index: 1.4360
Odor: Recognizable
Melting Point: -91°
Boiling Point: 87-89°
Beilstein Reference: 635697
UN No.: 1993
MDL No.: MFCD00001307
Formula:
C₄H₇Cl Boiling Pt: 87…89 °C Melting Pt: –91 °C Density: 0.980 Flash Pt: –1 °C (30 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001307 CAS Number: 5911-08-0 EINECS: 227-632-1 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...