Benzoyl isothiocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger

L06964-06 L06964-14
CAAAAL06964-06EA 71.42 CAD
CAAAAL06964-06 CAAAAL06964-14
Benzoyl isothiocyanate 98%
Benzoyl isothiocyanate

Purity: 98% CAS No.: 532-55-8 Molecular Formula: C8H5NOS Formula Weight: 163.20 Flash Point: 110°(230°F) Refractive Index: 1.6375 Odor: Pungent Boiling Point: 128-131°/15mm Beilstein Reference: 606716 Merck Reference: 14,1114 UN No.: 2922 MDL No.: MFCD00004815

Formula: C₈H₅NOS
MW: 163.2 g/mol
Boiling Pt: 252
Melting Pt: -1
Density: 1.214
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004815
CAS Number: 532-55-8
EINECS: 208-540-0
UN: 2810
ADR: 6.1 III
Merck Index: 14,1114

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR