2-Chloroanisole 98%
Supplier: Thermo Scientific Chemicals
Warning
|
L06952-18
L06952-30
CAAAAL06952-18EA
90.53
CAD
CAAAAL06952-18
CAAAAL06952-30
2-Chloroanisole 98%
2-Chloroanisole
Purity: 98%
CAS No.: 766-51-8
Molecular Formula: C7H7ClO
Formula Weight: 142.59
Flash Point: 76°(168°F)
Refractive Index: 1.5450
Boiling Point: 194-196°
Beilstein Reference: 2042179
MDL No.: MFCD00000557
Formula:
C₇H₇ClO Boiling Pt: 194…196 °C Melting Pt: –27 °C Density: 1,123 Flash Pt: 76 °C(168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000557 CAS Number: 766-51-8 EINECS: 212-167-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...