5-Methylindole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06893-03
L06893-06
CAAAAL06893-03EA
102.54
CAD
CAAAAL06893-03
CAAAAL06893-06
5-Methylindole 99%
5-Methylindole
Purity: 99%
CAS No.: 614-96-0
Molecular Formula: C9H9N
Formula Weight: 131.18
Storage Temperature: 2°C to 8°C
Melting Point: 58-61°
Boiling Point: 100°/3mm
Beilstein Reference: 2228
MDL No.: MFCD00005680
Formula:
C₉H₉N Boiling Pt: 100 °C/3 mm Melting Pt: 58…61 °C Density: 1.0202 g/cm³ (78.3 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005680 CAS Number: 614-96-0 EINECS: 210-400-9 UN: 3335 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...