2-Amino-6-chlorobenzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06883-03
L06883-06
CAAAAL06883-03EA
41.87
CAD
CAAAAL06883-03
CAAAAL06883-06
2-Amino-6-chlorobenzonitrile 98%
2-Amino-6-chlorobenzonitrile
Purity: 98%
CAS No.: 6575-11-7
Molecular Formula: C7H5ClN2
Formula Weight: 152.58
Melting Point: 133-137°
Beilstein Reference: 2717090
UN No.: 3439
MDL No.: MFCD00015546
Formula:
C₇H₅ClN₂ Boiling Pt: 301 °C (1013 hPa) Melting Pt: 133…137 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015546 CAS Number: 6575-11-7 EINECS: 229-501-4 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...