2,2,2-Trichloroethyl chloroformate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L06875-14
L06875-22
CAAAAL06875-14EA
83.9
CAD
CAAAAL06875-14
CAAAAL06875-22
2,2,2-Trichloroethyl chloroformate 97%
2,2,2-Trichloroethyl chloroformate
Purity: 97%
CAS No.: 17341-93-4
Molecular Formula: ClCO2CH2CCl3
Formula Weight: 211.86
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₃H₂Cl₄O₂ Boiling Pt: 171…172 °C Melting Pt: 0…2 °C Density: 1.539 Flash Pt: >100 °C (212 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000810 CAS Number: 17341-93-4 EINECS: 241-363-7 UN: 3277 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...