(±)-1,2-Hexanediol 97%

Supplier: Thermo Scientific Chemicals
Warning

L06864-18 L06864-30
CAAAAL06864-18EA 92.11 CAD
CAAAAL06864-18 CAAAAL06864-30
(±)-1,2-Hexanediol 97%
(±)-1,2-Hexanediol

Purity: 97% CAS No.: 6920-22-5 Molecular Formula: C6H14O2 Formula Weight: 118.18 Flash Point: 122°(251°F) Refractive Index: 1.4420 Boiling Point: 223-224° Beilstein Reference: 1719244 MDL No.: MFCD00010737

Formula: C₆H₁₄O₂
Boiling Pt: 223…224 °C
Melting Pt: 45 °C
Density: 0,951
Flash Pt: 122 °C (251 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010737
CAS Number: 6920-22-5
EINECS: 230-029-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR