Disoprofol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L06841-14
L06841-22
L06841-36
CAAAAL06841-14EA
49.45
CAD
CAAAAL06841-14
CAAAAL06841-22
CAAAAL06841-36
Disoprofol 97%
Disoprofol
An anesthetic that exhibits direct activation of the GABA-A receptor
Purity: 97%
CAS No.: 2078-54-8
Molecular Formula: C12H18O
Formula Weight: 178.28
Flash Point: 113°(235°F)
Density: 0.962
Refractive Index: 1.5140
Melting Point: 17-18°
Boiling Point: 254-256°
Beilstein Reference: 1866484
Merck Reference: 14,7834
MDL No.: MFCD00008885
An anesthetic that exhibits direct activation of the GABA-A receptor
Formula:
C₁₂H₁₈O Boiling Pt: 254…256 °C Melting Pt: 17…18 °C Density: 0.962 Flash Pt: 113 °C (235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008885 CAS Number: 2078-54-8 EINECS: 218-206-6 UN: 3145 ADR: 8,III Merck Index: 14,7834 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...