(±)-Ethyl-2-methylbutyrate 98%
Supplier: Thermo Scientific Chemicals
L06743-AE
L06743-AP
CAAAAL06743-AEEA
69.84
CAD
CAAAAL06743-AE
CAAAAL06743-AP
(±)-Ethyl-2-methylbutyrate 98%
(±)-Ethyl 2-methylbutyrate
Purity: 98%
CAS No.: 7452-79-1
Molecular Formula: C7H14O2
Formula Weight: 130.19
Flash Point: 26°(78°F)
Refractive Index: 1.3970
Boiling Point: 132-133°
Beilstein Reference: 1720887
UN No.: 3272
MDL No.: MFCD00012217
Formula:
C₇H₁₄O₂ |
MDL Number:
MFCD00012217 CAS Number: 7452-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...