1-Phenyl-2-thiourea 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Phenylthiourea, 1-Phenylthiourea

L06690-09 L06690-18 L06690-30
CAAAAL06690-09EA 73.94 CAD
CAAAAL06690-09 CAAAAL06690-18 CAAAAL06690-30
1-Phenyl-2-thiourea 97%
1-Phenyl-2-thiourea

Purity: 97% CAS No.: 103-85-5 Molecular Formula: C7H8N2S Formula Weight: 152.21 Odor: Odorless Melting Point: 149-153° Beilstein Reference: 907309 Merck Reference: 14,7315 UN No.: 2811 MDL No.: MFCD00004933

Formula: C₇H₈N₂S
MW: 152.22 g/mol
Boiling Pt: 295 - 297 °C (1013 hPa)
Melting Pt: 149…153 °C
Density: 1.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004933
CAS Number: 103-85-5
EINECS: 203-151-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,7315

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR