1,6-Hexanedithiol 97%
Supplier: Thermo Scientific Chemicals
Warning
|
L06686-06
L06686-14
CAAAAL06686-06EA
45.5
CAD
CAAAAL06686-06
CAAAAL06686-14
1,6-Hexanedithiol 97%
1,6-Hexanedithiol
Purity: 97%
CAS No.: 1191-43-1
Molecular Formula: HS(CH2)6SH
Formula Weight: 150.30
Flash Point: 90°(194°F)
Refractive Index: 1.5130
Melting Point: -21°
Boiling Point: 242-243°
Beilstein Reference: 1732507
UN No.: 2810
MDL No.: MFCD00004910
Formula:
C₆H₁₄S₂ Boiling Pt: 242…243 °C Melting Pt: –21 °C Density: 0.990 Flash Pt: 90 °C(194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004910 CAS Number: 1191-43-1 EINECS: 214-735-1 UN: 3334 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...