5-Ethyl-2-furaldehyde 98%
Supplier: Thermo Scientific Chemicals
Warning
|
L06659-03
L06659-06
CAAAAL06659-03EA
67.62
CAD
CAAAAL06659-03
CAAAAL06659-06
5-Ethyl-2-furaldehyde 98%
5-Ethyl-2-furaldehyde
Purity: 98%
CAS No.: 23074-10-4
Molecular Formula: C7H8O2
Formula Weight: 124.14
Flash Point: 65°(149°F)
Refractive Index: 1.5220
Boiling Point: 79-81°/12mm
Beilstein Reference: 1982
MDL No.: MFCD00030605
Formula:
C₇H₈O₂ Boiling Pt: 79…81 °C/12 mm Density: 1.055 Flash Pt: 65 °C(149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00030605 CAS Number: 23074-10-4 EINECS: 245-411-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...