3,4-Hexanedione 94%

Supplier: Thermo Scientific Chemicals
Warning

L06655-14 L06655-22
CAAAAL06655-14EA 77.42 CAD
CAAAAL06655-14 CAAAAL06655-22
3,4-Hexanedione 94%
3,4-Hexanedione

Purity: 94% CAS No.: 4437-51-8 Molecular Formula: C6H10O2 Formula Weight: 114.15 Flash Point: 29°(84°F) Refractive Index: 1.4120 Melting Point: -10° Boiling Point: 129-131° Beilstein Reference: 1700837 UN No.: 1224 MDL No.: MFCD00010237

Formula: C₆H₁₀O₂
Boiling Pt: 129…131 °C
Melting Pt: –10 °C
Density: 0.941
Flash Pt: 29 °C (84 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010237
CAS Number: 4437-51-8
EINECS: 224-651-7
UN: 1224
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR