Benzyl cyanoacetate 97%

Supplier: Thermo Scientific Chemicals
Warning

L06644-14 L06644-22
CAAAAL06644-14EA 126.24 CAD
CAAAAL06644-14 CAAAAL06644-22
Benzyl cyanoacetate 97%
Benzyl cyanoacetate

Purity: 97% CAS No.: 14447-18-8 Molecular Formula: NCCH2CO2CH2C6H5 Formula Weight: 175.18 Flash Point: 100°(212°F) Refractive Index: 1.5195 Odor: Odorless Melting Point: 26-30° Boiling Point: 140°/0.8mm Beilstein Reference: 2096578 MDL No.: MFCD00051719

Formula: C₁₀H₉NO₂
Boiling Pt: 140 °C (0.8 mmHg)
Melting Pt: 26…30 °C
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051719
CAS Number: 14447-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR