(±)-trans-1,2-Diaminocyclohexane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(±)-trans-1,2-Cyclohexanediamine, trans-Cyclohexane-1,2-diamine
L06626-09
L06626-18
L06626-30
CAAAAL06626-09EA
74.58
CAD
CAAAAL06626-09
CAAAAL06626-18
CAAAAL06626-30
(±)-trans-1,2-Diaminocyclohexane 99%
(±)-trans-1,2-Diaminocyclohexane
Purity: 99%
CAS No.: 1121-22-8
Molecular Formula: C6H14N2
Formula Weight: 114.19
Storage Temperature: 2°C to 8°C
Flash Point: 68°(154°F)
Refractive Index: 1.4890
Odor: Amine-like
Melting Point: 14-15°
Boiling Point: 89-90°/14mm
Beilstein Reference: 3193807
UN No.: 2735
MDL No.: MFCD00063747
Formula:
C₆H₁₄N₂ MW: 114.19 g/mol Boiling Pt: 89…90 °C/14 mm Melting Pt: 14…15 °C Density: 0.951 Flash Pt: 68 °C (154 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00063747 CAS Number: 1121-22-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...