2-Chloro-5-nitrobenzotrifluoride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-5-nitrobenzotrifluoride, 2-Chloro-α,α,α-trifluoro-5-nitrotoluene, 1-Chloro-4-nitro-2-(trifluoromethyl)benzene

L06623-14 L06623-22
CAAAAL06623-14EA 63.36 CAD
CAAAAL06623-14 CAAAAL06623-22
2-Chloro-5-nitrobenzotrifluoride 98%
2-Chloro-5-nitrobenzotrifluoride

Purity: 98% CAS No.: 777-37-7 Molecular Formula: C7H3ClF3NO2 Formula Weight: 225.55 Flash Point: 98°(208°F) Refractive Index: 1.5080 Melting Point: ca 22° Boiling Point: 232° Beilstein Reference: 2216169 MDL No.: MFCD00007296

Formula: C₇H₃ClF₃NO₂
MW: 225.55 g/mol
Boiling Pt: 232 °C
Melting Pt: ∼ 22 °C
Density: 1.547
Flash Pt: 98 °C (208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007296
CAS Number: 777-37-7
EINECS: 212-287-1
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR