6-Nitro-m-toluic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Nitro-5-methylbenzoic acid, 5-Methyl-2-nitrobenzoic acid

L06594-06 L06594-14
CAAAAL06594-06EA 72.05 CAD
CAAAAL06594-06 CAAAAL06594-14
6-Nitro-m-toluic acid 98+%
6-Nitro-m-toluic acid

Purity: 98+% CAS No.: 3113-72-2 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Melting Point: 133-137° Beilstein Reference: 1959031

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 364
Melting Pt: 133…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00007368
CAS Number: 3113-72-2
EINECS: 221-481-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR